Assay Details
| Gene Symbol: Il17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005683.1 | - | 1 - 2 | 142 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Mir143 and Mir145 host gene (non-coding RNA) | Mir143hg |
E330013P06, Gm16908 |
-25.5kb | Sense | Chr. 18 - 61645878 - 61665538 on NCBI Build GRCm38 |
| Downstream | prenylcysteine oxidase 1 like | Pcyox1l |
BC060677, C630049M13 |
3.8kb | Sense | Chr. 18 - 61696164 - 61707635 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Il17b
Genomic Map