Assay Details
Gene Symbol: 1700011L03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005875.1 | - | 1 - 2 | 144 | 61 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BY706018.1 | - | 148 | Chr. - - [-] on Build | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | interferon regulatory factor 2 | Irf2 |
9830146E22Rik, AI646973, Irf-2 |
-99.3kb | Anti-sense | Chr. 8 - 46739745 - 46847458 on NCBI Build GRCm38 |
Downstream | storkhead box 2 | Stox2 |
4933409N07Rik, AI449080, AI835114, mKIAA1392 |
231.2kb | Sense | Chr. 8 - 47180048 - 47413081 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
