Assay Details
Gene Symbol: Med9os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005951.1 | - | 1 - 2 | 86 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1810063I02 gene | 1810063I02Rik | -25.0kb | Anti-sense | Chr. 11 - 59921499 - 59921896 on NCBI Build GRCm38 | |
Downstream | mediator complex subunit 9 | Med9 |
BC019367, Med25 |
123.0 | Anti-sense | Chr. 11 - 59948214 - 59963306 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
