Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1430325_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1431264_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430325_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430325_at |
- |
- |
Mouse Genome 430 2.0 Array |
1431264_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200013835.mm.1 |
NR_038349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013835.mm.1 |
NR_038349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013835.mm.1 |
NR_038350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013835.mm.1 |
NR_038350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013835.mm.1 |
TC0200001799.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013840.mm.1 |
NR_038350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013840.mm.1 |
NR_038350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013840.mm.1 |
TC0200001799.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013841.mm.1 |
NR_038349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013841.mm.1 |
NR_038349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013841.mm.1 |
NR_038350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013841.mm.1 |
NR_038350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013841.mm.1 |
TC0200001799.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1430325_at |
- |
- |
Mouse Expression Set 430 |
1431264_at |
- |
- |
Mouse Expression Set 430 |
1431264_PM_at |
- |
- |
Mouse Expression Set 430 |
1430325_PM_at |
- |
- |
Murine Genome U74v2 Set |
167485_PM_at |
- |
- |
Murine Genome U74v2 Set |
167008_PM_i_at |
- |
- |
Murine Genome U74v2 Set |
167485_at |
- |
- |
Murine Genome U74v2 Set |
167008_i_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1430325_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1430325_at |
- |
- |