Assay Details
| Gene Symbol: 1700029B24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006483.1 | - | 1 | 347 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fatty acid binding protein 4, adipocyte | Fabp4 | 422/aP2, ALBP/Ap2, Ap2, Lbpl | -33.2kb | Anti-sense | Chr. 3 - 10204342 - 10208597 on NCBI Build GRCm38 | 
| Downstream | fatty acid binding protein 12 | Fabp12 | 1700008G05Rik | 1.9kb | Anti-sense | Chr. 3 - 10244209 - 10301526 on NCBI Build GRCm38 | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Amplicon length between 86 and 100  3' Most  Detect Genomic DNA  Mus musculus  Both primers and probe map within a single exon | 
MicroArray Probes:
 Back To Top
 Back To Top
 1700029B24Rik
1700029B24Rik Genomic Map
Genomic Map Back To Top
 Back To Top