Assay Details
| Gene Symbol: 1700056E22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028516.1 | NP_082792.1 | 1 | 858 | 104 | |
| GenBank mRNA | AK006811.1 | - | 1 | 442 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 34068 |
Gm34068 | -223.3kb | Anti-sense | Chr. 1 - 183748977 - 183809746 on NCBI Build GRCm38 | |
| Downstream | dual specificity phosphatase 10 | Dusp10 |
2610306G15Rik, AI158871, MKP-5, MKP5 |
887.0 | Anti-sense | Chr. 1 - 184034461 - 184075636 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700056E22Rik
Genomic Map