Assay Details
| Gene Symbol: 1700057A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK006821.1 | - | 1 | 355 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 171, member A1 |
Fam171a1 |
9630050M13Rik, AA409679 |
Sense | Chr. 2 - 3114202 - 3227809 on NCBI Build GRCm38 | |
| Downstream | N-myristoyltransferase 2 | Nmt2 |
A930001K02Rik, AI605445, AU044698, hNMT-2 |
135.0kb | Sense | Chr. 2 - 3284212 - 3328877 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700057A11Rik
Genomic Map