Assay Details
Gene Symbol: 1700084F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045965.1 | 4 - 5 | 497 | 86 | ||
GenBank mRNA | AK006991.1 | - | 4 - 5 | 498 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4933416O17 gene | 4933416O17Rik | -42.0kb | Anti-sense | Chr. 13 - 69957962 - 69962084 on NCBI Build GRCm38 | |
Downstream | predicted gene 3772 | Gm3772 | 1.1kb | Anti-sense | Chr. 13 - 70021959 - 70125917 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2416837 | AK006991.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774803 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459582 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398104 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763471 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868250 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592039 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52869 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111637 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02714483 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02714483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119131 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119107 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10452085 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119033 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10834018 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119129 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119016 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119049 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29118991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119117 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29118993 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119135 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119008 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119012 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10834017 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10908113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119134 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119041 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119010 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10555220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10944311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119017 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29118989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119015 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119037 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119006 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119106 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10555221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10959883 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29119120 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1430508_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430508_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430508_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300014907.mm.1 | NR_045965.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014907.mm.1 | NR_045965.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300014907.mm.1 | TC1300002282.mm.1 | MTA TC |
Mouse Expression Set 430 | 1430508_PM_at | - | - |
Mouse Expression Set 430 | 1430508_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430508_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430508_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106940086 | - | - |
