Assay Details
Gene Symbol: 1700096K18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027388.1 | 1 | 446 | 110 | ||
GenBank mRNA | AK007094.1 | - | 1 | 452 | 110 | |
AK047471.1 | - | 1 | 447 | 110 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
chaperonin containing TCP1, subunit 8 (theta)-like 1 |
Cct8l1 | Gm443 | -12.0kb | Sense | Chr. 5 - 25516067 - 25518027 on NCBI Build GRCm38 |
Downstream | X-ray repair complementing defective repair in Chinese hamster cells 2 | Xrcc2 |
4921524O04Rik, 8030409M04Rik, RAD51, RecA |
159.0kb | Anti-sense | Chr. 5 - 25689814 - 25705797 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
