Assay Details
| Gene Symbol: 1700125C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007275.1 | - | 1 | 249 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 38, member 9 |
Slc38a9 |
4833412L08Rik, 6720411P22Rik, 9130023D20Rik, 9430067K09Rik, A730092C09 |
-61.6kb | Anti-sense | Chr. 13 - 112660691 - 112738752 on NCBI Build GRCm38 |
| Downstream | superkiller viralicidic activity 2-like 2 (S. cerevisiae) | Skiv2l2 |
2610528A15Rik, mKIAA0052 |
67.1kb | Sense | Chr. 13 - 112867780 - 112927380 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700125C05Rik
Genomic Map