Assay Details
| Gene Symbol: Zim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007312.2 | - | 201 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 264 | Zfp264 | Anti-sense | Chr. 7 - 6977413 - 6985640 on NCBI Build GRCm38 | ||
| Downstream | aurora kinase C | Aurkc |
AIE1, AIK3, ARK-3, IAK3, Stk13 |
12.5kb | Anti-sense | Chr. 7 - 6989892 - 7003364 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Zim3
Genomic Map