Assay Details
Gene Symbol: 1810057I12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK007886.1 | - | 1 | 249 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glucagon-like peptide 1 receptor | Glp1r |
GLP-1R, GLP1Rc |
-492.0 | Sense | Chr. 17 - 30901867 - 30936510 on NCBI Build GRCm38 |
Downstream | uromodulin-like 1 | Umodl1 | D17Ertd488e | 17.1kb | Sense | Chr. 17 - 30954640 - 31010710 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
