Assay Details
Gene Symbol: 2010109N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK008001.1 | - | 1 - 2 | 120 | 85 | |
AK008371.1 | - | 1 - 2 | 120 | 85 | ||
AK008488.1 | - | 1 - 2 | 120 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MARVEL (membrane-associating) domain containing 3 | Marveld3 |
1810006A16Rik, AI642133, Marvd3, Mrvldc3 |
-23.8kb | Anti-sense | Chr. 8 - 109947909 - 109962205 on NCBI Build GRCm38 |
Downstream | tyrosine aminotransferase | Tat | 3.7kb | Sense | Chr. 8 - 109990436 - 109999804 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
