Assay Details
Gene Symbol: 2310001K20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK009069.1 | - | 1 | 154 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATP/GTP binding protein-like 1 | Agbl1 |
Ccp4, D430020F16, EG244071, Nna1-l1 |
Sense | Chr. 7 - 76229887 - 77124698 on NCBI Build GRCm38 | |
Downstream |
neurotrophic tyrosine kinase, receptor, type 3 |
Ntrk3 |
AW125844, Ntrk3_tv3, TrkC |
1.6mb | Anti-sense | Chr. 7 - 78179986 - 78578852 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
