Assay Details
Gene Symbol: 2310016D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045491.1 | 2 - 3 | 81 | |||
GenBank mRNA | AK009380.1 | - | 2 - 3 | 160 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
syntrophin, gamma 2 |
Sntg2 |
2210008K22Rik, 9530013L23, BB121248 |
-37.2kb | Sense | Chr. 12 - 30174480 - 30373392 on NCBI Build GRCm38 |
Downstream | transmembrane protein 18 | Tmem18 | BC024093 | 156.1kb | Anti-sense | Chr. 12 - 30584443 - 30591219 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
