Assay Details
| Gene Symbol: 4933406J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020838.1 | - | 1 | 545 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | elongation factor Tu GTP binding domain containing 1 | Eftud1 |
4932434J20Rik, 6030468D11Rik, AI451340, AU019507, AU022896, D7Ertd791e |
Anti-sense | Chr. 7 - 82648614 - 82777852 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 29796 |
Gm29796 | 56.0kb | Anti-sense | Chr. 7 - 82810903 - 82823444 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4933406J10Rik
Genomic Map