Assay Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020945.1 | - | 1 | 953 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 16630 |
Gm16630 | -23.9kb | Anti-sense | Chr. 6 - 48128658 - 48395408 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein 467 | Zfp467 |
1190001I08Rik, AB041616, AI426952, AV017623, EZI, MNCb-3350, Znf467 |
7.3kb | Anti-sense | Chr. 6 - 48427692 - 48445998 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Krba1
Genomic Map