Assay Details
| Gene Symbol: B230112G18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020967.1 | - | 1 | 444 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta) |
Agpat6 |
AU041707, AW545732, GPAT4, Tsarg7 |
Anti-sense | Chr. 8 - 23171265 - 23209171 on NCBI Build GRCm38 | |
| Downstream | GINS complex subunit 4 (Sld5 homolog) | Gins4 |
2810037C03Rik, 4933405K01Rik, Sld5 |
17.2kb | Anti-sense | Chr. 8 - 23226610 - 23237668 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

B230112G18Rik
Genomic Map