Assay Details
| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021139.1 | - | 3 - 4 | 402 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ZXD family zinc finger C | Zxdc |
A930012H20, B930086F11Rik, BC003332 |
-17.5kb | Anti-sense | Chr. 6 - 90369483 - 90403490 on NCBI Build GRCm38 |
| Downstream | Kruppel-like factor 15 | Klf15 |
1810013I09Rik, AV048136, AW494632, CKLF, KKLF |
41.2kb | Anti-sense | Chr. 6 - 90462626 - 90479170 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Ccdc37
Genomic Map