Assay Details
| Gene Symbol: C030045M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021151.1 | - | 1 | 953 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
diacylglycerol kinase, beta |
Dgkb |
6430574F24, 90kda, C630029D13Rik, DAGK2, DGK, DGK-beta, mKIAA0718 |
Sense | Chr. 12 - 37880168 - 38634239 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930428E07 gene | 4930428E07Rik | 50.2kb | Anti-sense | Chr. 12 - 38552809 - 38614521 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C030045M22Rik
Genomic Map