Assay Details
| Gene Symbol: C430014O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021240.1 | - | 1 | 548 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
dyskeratosis congenita 1, dyskerin homolog pseudogene |
Gm19052 | -90.3kb | Sense | Chr. 18 - 10236687 - 10237622 on NCBI Build GRCm38 | |
| Downstream | establishment of cohesion 1 homolog 1 (S. cerevisiae) | Esco1 | A930014I12Rik | 237.7kb | Anti-sense | Chr. 18 - 10566507 - 10610685 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C430014O12Rik
Genomic Map