Assay Details
| Gene Symbol: C430045I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021252.1 | - | 1 | 1055 | 71 | |
| BG066234.2 | - | 1 | 71 | |||
| C78534.1 | - | 1 | 71 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein kinase C and casein kinase substrate in neurons 2 | Pacsin2 | AI197433 | Sense | Chr. 15 - 83375607 - 83464606 on NCBI Build GRCm38 | |
| Downstream | tubulin tyrosine ligase-like 1 | Ttll1 |
6330444E16Rik, AV014541 |
84.9kb | Sense | Chr. 15 - 83483769 - 83518917 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C430045I18Rik
Genomic Map