Assay Details
Gene Symbol: C630043D15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021277.1 | - | 1 | 448 | 105 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3 | St8sia3 |
AI847333, ST8SiaIII, Siat8c |
Sense | Chr. 18 - 64254359 - 64276144 on NCBI Build GRCm38 | |
Downstream |
one cut domain, family member 2 |
Onecut2 |
C730009D12, OC-2, Oc2 |
72.0kb | Sense | Chr. 18 - 64340364 - 64398488 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
