Assay Details
| Gene Symbol: D530043N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021315.1 | - | 1 | 353 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13964 | Gm13964 | OTTMUSG00000015197 | -146.4kb | Anti-sense | Chr. 2 - 113499946 - 113504071 on NCBI Build GRCm38 | 
| Downstream | RIKEN cDNA 4930533B01 gene | 4930533B01Rik | 40.1kb | Anti-sense | Chr. 2 - 113691053 - 113708550 on NCBI Build GRCm38 | 
More Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			Set Membership: | 
		   
					
						
							 | 
				

D530043N20Rik
Genomic Map