Assay Details
Gene Symbol: St6galnac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021356.1 | - | 1 | 653 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700012D16 gene | 1700012D16Rik | -210.2kb | Sense | Chr. 3 - 153296502 - 153298649 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 5730460C07 gene | 5730460C07Rik | 279.6kb | Sense | Chr. 3 - 153789305 - 153792087 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
