Assay Details
Gene Symbol: E130114A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021388.1 | - | 1 | 846 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glycerol kinase | Gyk |
D930012N15Rik, GK |
Sense | Chr. X - 85701937 - 85776819 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 5430427O19 gene | 5430427O19Rik | 42.2kb | Anti-sense | Chr. X - 85776957 - 85891499 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
