Assay Details
Gene Symbol: C030015A19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028820.1 | - | 1 | 2248 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | urocanase domain containing 1 | Uroc1 |
AI265605, BC022133 |
-22.3kb | Sense | Chr. 6 - 90333239 - 90364551 on NCBI Build GRCm38 |
Downstream | coiled-coil domain containing 37 | Ccdc37 |
AW048498, C030041G11Rik, C230069K22Rik |
14.4kb | Anti-sense | Chr. 6 - 90403731 - 90430246 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
