Assay Details
Gene Symbol: D730044K07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029389.1 | - | 1 | 1652 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ribosomal protein L30 | Rpl30 | Anti-sense | Chr. 15 - 34440505 - 34443276 on NCBI Build GRCm38 | ||
Downstream | glutamate rich 5 | Erich5 | 8.5kb | Sense | Chr. 15 - 34453312 - 34473892 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
