Assay Details
| Gene Symbol: 4932416H05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029452.1 | 1 | 2348 | 87 | ||
| GenBank mRNA | AK030036.1 | - | 1 | 2349 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prodynorphin | Pdyn | Dyn | -98.4kb | Sense | Chr. 2 - 129686549 - 129699938 on NCBI Build GRCm38 | 
| Downstream | transglutaminase 3, E polypeptide | Tgm3 | AI893889, TG E, TG(E), TGE, TGase E, TGase-3 | 186.3kb | Anti-sense | Chr. 2 - 129987794 - 130050399 on NCBI Build GRCm38 | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Amplicon length between 86 and 100  3' Most  Detect Genomic DNA  Mus musculus  1700  Both primers and probe map within a single exon | 
MicroArray Probes:
 Back To Top
 Back To Top
 4932416H05Rik
4932416H05Rik Genomic Map
Genomic Map Back To Top
 Back To Top