Assay Details
| Gene Symbol: Lemd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI849139.1 | - | 1 | 84 | ||
| AK032278.1 | - | 1 | 4047 | 84 | ||
| BC066109.1 | - | 2 | 5165 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 135b | Mir135b |
Mirn135b, mir-135b |
-7.2kb | Sense | Chr. 1 - 132198088 - 132198184 on NCBI Build GRCm38 |
| Downstream | kelch domain containing 8A | Klhdc8a | A630065K24Rik | 88.9kb | Sense | Chr. 1 - 132298490 - 132307357 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lemd1
Genomic Map