Assay Details
Gene Symbol: 9230105E05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040626.1 | 2 - 3 | 338 | 76 | ||
GenBank mRNA | AK033763.1 | - | 2 - 3 | 340 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
LLP homolog, long-term synaptic facilitation (Aplysia) |
Llph | 1190005P17Rik | -157.7kb | Sense | Chr. 10 - 120227060 - 120232071 on NCBI Build GRCm38 |
Downstream | microRNA 763 | Mir763 |
Mirn763, mir-763, mmu-mir-763 |
55.2kb | Anti-sense | Chr. 10 - 120447991 - 120448110 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
