Assay Details
Gene Symbol: AW549542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045702.1 | 4 - 5 | 1526 | 121 | ||
GenBank mRNA | AI413450.1 | - | 1 - 2 | 121 | ||
AK035112.1 | - | 1548 | 121 | |||
AK148622.1 | - | 1 - 2 | 225 | 121 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 31314 |
Gm31314 | -42.2kb | Anti-sense | Chr. 5 - 119472143 - 119527976 on NCBI Build GRCm38 | |
Downstream |
T-box 3, opposite strand 1 |
Tbx3os1 | Gm2301 | 27.2kb | Anti-sense | Chr. 5 - 119597977 - 119671331 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2274378 | AW549542.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2274378 | AK035112.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861163 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815183 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638314 | AB050614.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638314 | BC119343.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638314 | BC119345.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638314 | NM_032542.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718878 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96072 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918218 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206552 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766878 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957288 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618392 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173402 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312173 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626613 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73739 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174980 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338162 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90231 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171190 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580830 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40885 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140735 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12563 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151550 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127625 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2274378 | AW549542.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2274378 | AK035112.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05716919 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05716883 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127625 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01202197 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123182 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033468 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033587 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033469 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04819591 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04353454 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04499124 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04532678 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04532683 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04648774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033471 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04895174 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04873000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033593 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033592 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04438737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033401 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033605 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033609 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033614 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04338428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033472 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04408117 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033365 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033480 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04532679 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033606 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033373 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04581635 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04482299 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04406416 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033596 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04872998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033397 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033630 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033379 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033598 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033594 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033386 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033703 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04820613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033402 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04499118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04831551 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033425 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033586 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033698 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033396 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04273898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033700 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04532685 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033422 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033421 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033589 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24033382 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0500027969.mm.1 | TC0500003157.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500027969.mm.1 | NR_045702.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500027969.mm.1 | NR_045702.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500027970.mm.1 | TC0500003157.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500027970.mm.1 | NR_045702.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500027970.mm.1 | NR_045702.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500027971.mm.1 | TC0500003157.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500027971.mm.1 | NR_045702.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500027971.mm.1 | NR_045702.1 | Refseq |
Murine Genome U74v2 Set | 167473_r_at | - | - |
Murine Genome U74v2 Set | 167473_PM_r_at | - | - |
Murine Genome U74v2 Set | 130855_PM_at | - | - |
Murine Genome U74v2 Set | 130855_at | - | - |
