Assay Details
Gene Symbol: 9530085L11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035677.1 | - | 1 | 1553 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ADP-ribosylhydrolase like 1 | Adprhl1 |
Arh2, D330008N11Rik |
-31.1kb | Sense | Chr. 8 - 13235632 - 13254263 on NCBI Build GRCm38 |
Downstream | transmembrane and coiled-coil domains 3 | Tmco3 |
B230339H12Rik, C87304 |
783.0 | Anti-sense | Chr. 8 - 13288013 - 13322924 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
