Assay Details
| Gene Symbol: D030041H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037531.1 | - | 1 | 1253 | 92 | |
| AK083531.1 | - | 2 | 3088 | 92 | ||
| BB443518.2 | - | 1 | 327 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3075 | Mir3075 |
mir-3075, mmu-mir-3075 |
-18.3kb | Sense | Chr. 14 - 25534439 - 25534523 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4931406H21 gene | 4931406H21Rik | 32.4kb | Sense | Chr. 14 - 25586804 - 25590661 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D030041H20Rik
Genomic Map