Assay Details
| Gene Symbol: Ccl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038019.1 | - | 1 | 1247 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ELAV (embryonic lethal, abnormal vision)-like 1 (Hu antigen R) |
Elavl1 |
2410055N02Rik, HUR, Hua, W91709 |
-133.0 | Anti-sense | Chr. 8 - 4284781 - 4325100 on NCBI Build GRCm38 |
| Downstream | ceramide synthase 4 | Cers4 |
2900019C14Rik, Lass4, Trh1 |
165.3kb | Sense | Chr. 8 - 4492987 - 4526308 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccl25
Genomic Map