Assay Details
Gene Symbol: 2610307P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038887.1 | - | 1 - 2 | 39 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
prolactin family 5, subfamily a, member 1 |
Prl5a1 |
1600013P04Rik, D13Wsu14e, PLP-L, Prlpl |
-354.1kb | Anti-sense | Chr. 13 - 28142484 - 28151595 on NCBI Build GRCm38 |
Downstream | microRNA 6368 | Mir6368 |
Gm27800, mmu-mir-6368 |
189.7kb | Sense | Chr. 13 - 28710781 - 28710873 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
