Assay Details
| Gene Symbol: Pwwp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164231.1 | NP_001157703.1 | 4 | 3000 | 174 | |
| GenBank mRNA | AK039984.1 | - | 1 | 353 | 175 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
fatty acid binding protein 6, ileal (gastrotropin) |
Fabp6 |
GT, I-15P, I-BABP, ILBP, ILBP3, Illbp |
-119.6kb | Anti-sense | Chr. 11 - 43596040 - 43601562 on NCBI Build GRCm38 |
| Downstream | tetratricopeptide repeat domain 1 | Ttc1 |
4833412C19Rik, C79328, TPR1 |
7.8kb | Anti-sense | Chr. 11 - 43730006 - 43747973 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pwwp2a
Genomic Map