Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500021269.mm.1 |
TC0500002311.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500021269.mm.1 |
NR_028417.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500021269.mm.1 |
NR_028417.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500021267.mm.1 |
TC0500002311.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500021267.mm.1 |
NR_028417.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500021267.mm.1 |
NR_028417.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500021265.mm.1 |
TC0500002311.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500021265.mm.1 |
NR_028417.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500021265.mm.1 |
NR_028417.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500021266.mm.1 |
TC0500002311.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500021266.mm.1 |
NR_028417.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500021266.mm.1 |
NR_028417.1 |
Refseq |