Assay Details
| Gene Symbol: 1700028E10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045699.1 | 3 - 4 | 395 | 80 | ||
| GenBank mRNA | AK042042.1 | - | 3 - 4 | 362 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | StAR-related lipid transfer (START) domain containing 13 | Stard13 |
DLC2, GT650 |
Anti-sense | Chr. 5 - 151037515 - 151428256 on NCBI Build GRCm38 | |
| Downstream | predicted gene 3704 | Gm3704 | 27.8kb | Sense | Chr. 5 - 151423987 - 151432127 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2354296 | AK042042.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069582 | AK031407.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155453 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141252 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069582 | AK031407.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P663040 | AK031407.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P643893 | AK006454.1 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04943315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04379727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04524172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04524170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04939755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04939752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04939750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04901935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04889855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04495339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04939754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04524171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04379728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04939761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24196394 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1446606_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440352_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440352_at | - | - |
| Mouse Genome 430 2.0 Array | 1440352_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446606_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446606_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500017311.mm.1 | TC0500001878.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017311.mm.1 | NR_045699.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017311.mm.1 | NR_045699.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017307.mm.1 | TC0500001878.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017307.mm.1 | NR_045699.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017307.mm.1 | NR_045699.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017307.mm.1 | NR_045700.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017307.mm.1 | NR_045700.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017308.mm.1 | TC0500001878.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017308.mm.1 | NR_045699.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017308.mm.1 | NR_045699.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017308.mm.1 | NR_045700.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017308.mm.1 | NR_045700.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017310.mm.1 | TC0500001878.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017310.mm.1 | NR_045699.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017310.mm.1 | NR_045699.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017304.mm.1 | TC0500001878.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500017304.mm.1 | NR_045699.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017304.mm.1 | NR_045699.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017304.mm.1 | NR_045700.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500017304.mm.1 | NR_045700.1 | Refseq |
| Mouse Expression Set 430 | 1446606_PM_at | - | - |
| Mouse Expression Set 430 | 1440352_at | - | - |
| Mouse Expression Set 430 | 1446606_at | - | - |
| Mouse Expression Set 430 | 1440352_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106370242 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2497906 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2551437 | - | - |

1700028E10Rik
Genomic Map