Assay Details
| Gene Symbol: Gm13497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040636.1 | 1 - 2 | 389 | 94 | ||
| GenBank mRNA | AK042686.1 | - | 1 - 2 | 390 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho family GTPase 3 | Rnd3 |
2610017M01Rik, AI661404, Arhe, Rhoe |
-92.7kb | Anti-sense | Chr. 2 - 51130439 - 51149111 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700057H21 gene | 1700057H21Rik | 127.1kb | Anti-sense | Chr. 2 - 51421050 - 51437900 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm13497
Genomic Map