Assay Details
Gene Symbol: Anp32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK043419.1 | - | 1 | 2748 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hemogen | Hemgn |
4921524M03Rik, AI317176, EDAG, Hgn, mNDR |
-44.4kb | Anti-sense | Chr. 4 - 46393989 - 46413504 on NCBI Build GRCm38 |
Downstream | N-acetylneuraminic acid synthase (sialic acid synthase) | Nans |
4632418E04Rik, BB056474, Sas |
28.4kb | Sense | Chr. 4 - 46489329 - 46503438 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
