Assay Details
Gene Symbol: A830019L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040551.1 | 3 - 4 | 574 | 82 | ||
GenBank mRNA | AK043679.1 | - | 3 - 4 | 575 | 82 | |
AK086499.1 | - | 209 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 9530052C20 gene | 9530052C20Rik | -358.7kb | Sense | Chr. 3 - 142878399 - 142898015 on NCBI Build GRCm38 | |
Downstream | predicted gene 6260 | Gm6260 | EG621809 | 412.9kb | Anti-sense | Chr. 3 - 143670892 - 143712143 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746265 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686301 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652392 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954019 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170872 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43013 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41815 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05101362 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05101319 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129812 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05101319 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02296064 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02459136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595148 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595184 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595156 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02731780 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595151 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02296071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595183 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03188674 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595206 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03016090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595174 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595143 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595191 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595176 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02335639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595245 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02774295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595141 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595341 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595170 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595204 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595230 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595150 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595160 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595212 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595254 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02572789 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595274 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595352 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595328 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595181 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02335635 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02931605 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595216 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595153 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03188672 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595287 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595350 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02296062 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595203 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595171 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595223 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595166 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595313 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595275 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595279 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02774294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595172 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595195 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595146 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03188684 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02335634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595345 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595149 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02459139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22595334 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0300011745.mm.1 | NR_040551.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011745.mm.1 | NR_040551.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300011745.mm.1 | TC0300001455.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 413934 | - | - |
