Assay Details
Gene Symbol: Slc1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK046688.1 | - | 1 | 2355 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 31282 |
Gm31282 | -182.9kb | Anti-sense | Chr. 15 - 8468493 - 8502882 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 31340 |
Gm31340 | 22.8kb | Anti-sense | Chr. 15 - 8711026 - 8864483 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
