Assay Details
Gene Symbol: Apopt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK047131.1 | - | 1 | 2549 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tRNA methyltransferase 61A | Trmt61a |
6720458F09Rik, AI606093, Gcd14, Trm61 |
-28.9kb | Sense | Chr. 12 - 111678105 - 111683902 on NCBI Build GRCm38 |
Downstream | kinesin light chain 1 | Klc1 |
AI874768, Kns2 |
43.3kb | Sense | Chr. 12 - 111758849 - 111807844 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
