Assay Details
| Gene Symbol: B930046K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047296.1 | - | 1 | 2347 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NAD kinase | Nadk |
4432404C02Rik, BC004012 |
-8.3kb | Anti-sense | Chr. 4 - 155562392 - 155591001 on NCBI Build GRCm38 |
| Downstream | predicted gene 16023 | Gm16023 | OTTMUSG00000029160 | 6.5kb | Sense | Chr. 4 - 155608297 - 155624755 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

B930046K06Rik
Genomic Map