Assay Details
| Gene Symbol: C130079B09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK048576.1 | - | 1 | 2245 | 99 | |
| BB375259.2 | - | 1 | 469 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PTK2 protein tyrosine kinase 2 | Ptk2 |
FAK, FRNK, Fadk, mKIAA4203, p125FAK |
Sense | Chr. 15 - 73205102 - 73423275 on NCBI Build GRCm38 | |
| Downstream | DENN/MADD domain containing 3 | Dennd3 |
AI447457, E030003N15Rik |
109.4kb | Anti-sense | Chr. 15 - 73512560 - 73572242 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C130079B09Rik
Genomic Map