Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500017093.mm.1 |
NR_040423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017093.mm.1 |
NR_040423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017093.mm.1 |
TC1500002133.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017097.mm.1 |
NR_040423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017097.mm.1 |
NR_040423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017097.mm.1 |
TC1500002133.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017099.mm.1 |
NR_040423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017099.mm.1 |
NR_040423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017099.mm.1 |
TC1500002133.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017102.mm.1 |
NR_040423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017102.mm.1 |
NR_040423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017102.mm.1 |
TC1500002133.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017103.mm.1 |
NR_040423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017103.mm.1 |
NR_040423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017103.mm.1 |
TC1500002133.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017104.mm.1 |
NR_040423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017104.mm.1 |
NR_040423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017104.mm.1 |
TC1500002133.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017094.mm.1 |
NR_040423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017094.mm.1 |
NR_040423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017094.mm.1 |
TC1500002133.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017101.mm.1 |
NR_040423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017101.mm.1 |
NR_040423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017101.mm.1 |
TC1500002133.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500017095.mm.1 |
NR_040423.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017095.mm.1 |
NR_040423.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500017095.mm.1 |
TC1500002133.mm.1 |
MTA TC |