Assay Details
| Gene Symbol: 4930448O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015424.1 | - | 1 | 441 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 34388 |
Gm34388 | -86.8kb | Anti-sense | Chr. 13 - 97865811 - 97894001 on NCBI Build GRCm38 | |
| Downstream |
UTP15, U3 small nucleolar ribonucleoprotein, homolog (yeast) |
Utp15 | AW544865 | 265.4kb | Anti-sense | Chr. 13 - 98246845 - 98262992 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930448O17Rik
Genomic Map