Assay Details
| Gene Symbol: 4930519E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015840.1 | - | 1 | 1053 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 35191 |
Gm35191 | -1.3mb | Anti-sense | Chr. 5 - 39975676 - 39985628 on NCBI Build GRCm38 | |
| Downstream |
RAB28, member RAS oncogene family |
Rab28 |
2700023P08Rik, AW496496 |
342.1kb | Anti-sense | Chr. 5 - 41624974 - 41708192 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930519E07Rik
Genomic Map