Assay Details
| Gene Symbol: 4930524O05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045316.1 | 1 - 2 | 118 | 80 | ||
| GenBank mRNA | AK015890.1 | - | 1 - 2 | 119 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | damage specific DNA binding protein 1 | Ddb1 |
127kDa, AA408517, p127-Ddb1 |
-17.0kb | Anti-sense | Chr. 19 - 10605568 - 10629822 on NCBI Build GRCm38 |
| Downstream |
pepsinogen 5, group I |
Pga5 |
1110035E17Rik, Pepf |
20.9kb | Sense | Chr. 19 - 10668957 - 10678071 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930524O05Rik
Genomic Map